Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Vinylbenzeneboronic acid, 98%

$32.54 - $98.24

Chemical Identifiers

Chemical Name or Material 3-Vinylbenzeneboronic acid
CAS 15016-43-0
Molecular Formula C8H9BO2
Formula Weight 147.97
MDL Number MFCD01074679
Beilstein 3030426
Synonym Styrene-3-boronic acid; 3-Vinylphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1982903

  SDS
alfa aesar™
L1982903
1g Each for $98.24

AAL19829MD

  SDS
alfa aesar™
L19829MD
250mg Each for $32.54
Description & Specifications

Specifications

Formula Weight 147.97
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Chemical Name or Material 3-Vinylbenzeneboronic acid
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym Styrene-3-boronic acid; 3-Vinylphenylboronic acid
Assay 98%
MDL Number MFCD01074679
Molecular Formula C8H9BO2
Beilstein 3030426
Melting Point 127° to 129°C
Recommended Storage 2° to 8°C
CAS 15016-43-0
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C