Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Undecyne, 97%

$60.27 - $245.61

Chemical Identifiers

Chemical Name or Material 3-Undecyne
CAS 60212-30-8
Molecular Formula CH3(CH2)6C≡CCH2CH3
Formula Weight 152.28
MDL Number MFCD00041663
Synonym Ethyl n-heptyl acetylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2175306

  SDS
alfa aesar™
B2175306
5g Each for $60.27

AAB2175314

  SDS
alfa aesar™
B2175314
25g Each for $245.61
Description & Specifications

Specifications

Formula Weight 152.28
Assay 97%
Molecular Formula CH3(CH2)6C≡CCH2CH3
MDL Number MFCD00041663
CAS 60212-30-8
Synonym Ethyl n-heptyl acetylene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 79° to 81°C (10mmHg)
Quantity 5g
Chemical Name or Material 3-Undecyne
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.