missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Triphenylphosphoranylidene)tetrahydropyran-2-one, 97%, ACROS Organics™

Chemical Identifiers

CAS 218282-19-0
Molecular Formula C23H21O2P
Molecular Weight (g/mol) 360.39
InChI Key PZGUKMFSICDTKX-UHFFFAOYSA-N
Synonym 3-triphenyl-??-phosphanylidene oxan-2-one, 3-triphenylphosphoranylidene tetrahydropyran-2-one
PubChem CID 10594674
IUPAC Name 3-(triphenyl-$l^{5}-phosphanylidene)oxan-2-one
SMILES C1CC(=P(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)C(=O)OC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC440980010
SDS View Documents
Thermo Fisher Scientific
AC440980010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC440980050
SDS View Documents
Thermo Fisher Scientific
AC440980050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

218282-19-0
360.39
3-triphenyl-??-phosphanylidene oxan-2-one, 3-triphenylphosphoranylidene tetrahydropyran-2-one
3-(triphenyl-$l^{5}-phosphanylidene)oxan-2-one
C23H21O2P
PZGUKMFSICDTKX-UHFFFAOYSA-N
10594674
C1CC(=P(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)C(=O)OC1

Specifications

Glass bottle
1g
96% min. (HPLC)
3-triphenyl-??-phosphanylidene oxan-2-one, 3-triphenylphosphoranylidene tetrahydropyran-2-one
C1CC(=P(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)C(=O)OC1
360.39
360.39
3-(Triphenylphosphoranylidene)tetrahydropyran-2-one
228.0°C to 230.0°C
218282-19-0
C23H21O2P
PZGUKMFSICDTKX-UHFFFAOYSA-N
3-(triphenyl-$l^{5}-phosphanylidene)oxan-2-one
10594674
97%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS