missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Triphenylphosphoranylidene)tetrahydropyran-2-one, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC440980050
Specifications
3-(Triphenylphosphoranylidene)tetrahydropyran-2-one | |
228.0°C to 230.0°C | |
218282-19-0 | |
C23H21O2P | |
PZGUKMFSICDTKX-UHFFFAOYSA-N | |
3-(triphenyl-$l^{5}-phosphanylidene)oxan-2-one | |
10594674 | |
97% |
Glass bottle | |
5g | |
96% min. (HPLC) | |
3-triphenyl-??-phosphanylidene oxan-2-one, 3-triphenylphosphoranylidene tetrahydropyran-2-one | |
C1CC(=P(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)C(=O)OC1 | |
360.39 | |
360.39 |
Chemical Identifiers
218282-19-0 | |
360.39 | |
3-triphenyl-??-phosphanylidene oxan-2-one, 3-triphenylphosphoranylidene tetrahydropyran-2-one | |
3-(triphenyl-$l^{5}-phosphanylidene)oxan-2-one |
C23H21O2P | |
PZGUKMFSICDTKX-UHFFFAOYSA-N | |
10594674 | |
C1CC(=P(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)C(=O)OC1 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning