missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Trimethylsilylpropynal, 97%

Manufacturer:  Alfa Aesar™ 4476303

 View more versions of this product

Catalog No. AA4476303



Description

Description

  • Packaged under Argon
Specifications

Specifications

3-Trimethylsilylpropynal
29°C (84°F)
Air & Moisture Sensitive
Colorless to Yellow
2975-46-4
(CH3)3SiC≡CCHO
UN1993
LJRWLSKYGWLYIM-UHFFFAOYSA-N
3-trimethylsilylprop-2-ynal
2760865
Liquid
0.86g/mL
1.4435
58°C to 60°C (37.5mmHg)
1g
C6H10OSi
MFCD01103709
3-trimethylsilylpropynal, 3-trimethylsilyl-2-propynal, 3-trimethylsilyl propiolaldehyde, 3-trimethylsilyl propynal, trimethylsilyl propiolaldehyde, 2-propynal, 3-trimethylsilyl, trimethylsilylpropynal, 3-trimethylsilyl propynal, 3-trimethylsilyl-2-propynal, 3-trimethylsilyl prop-2-ynal
C[Si](C)(C)C#CC=O
126.23
126.23
97%

Chemical Identifiers

2975-46-4
126.23
LJRWLSKYGWLYIM-UHFFFAOYSA-N
2760865
C[Si](C)(C)C#CC=O
C6H10OSi
MFCD01103709
3-trimethylsilylpropynal, 3-trimethylsilyl-2-propynal, 3-trimethylsilyl propiolaldehyde, 3-trimethylsilyl propynal, trimethylsilyl propiolaldehyde, 2-propynal, 3-trimethylsilyl, trimethylsilylpropynal, 3-trimethylsilyl propynal, 3-trimethylsilyl-2-propynal, 3-trimethylsilyl prop-2-ynal
3-trimethylsilylprop-2-ynal
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

storageNote1 :

  • Packaged under Argon

Recommended Storage : 2°C to 8°C

SDS Documents