missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Trimethylsilylpropargyl aldehyde diethyl acetal 97+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC380930050

 View more versions of this product

Catalog No. AC380930050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

3-Trimethylsilylpropargyl aldehyde diethyl acetal
5g
97.0
Authentic
Liquid
97+%
(CH3)3SiCCCH(OCH2CH3)2
MFCD08275372
Clear colorless to slightly yellow
87219-80-5
100.0
1.4300 to 1.4320 (20°C, 589nm)
≥97.0%
C10H20O2Si
200.35

Chemical Identifiers

87219-80-5
100.0
C10H20O2Si
200.35
3-Trimethylsilylpropargyl aldehyde diethyl acetal
97.0
97+%
(CH3)3SiCCCH(OCH2CH3)2
MFCD08275372
≥97.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning