missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Trimethylsilylpropargyl aldehyde diethyl acetal 97+%, ACROS Organics™

Chemical Identifiers

CAS 87219-80-5
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Molecular Formula C10H20O2Si
Linear Formula (CH3)3SiCCCH(OCH2CH3)2
Formula Weight 200.35
MDL Number MFCD08275372
Chemical Name or Material 3-Trimethylsilylpropargyl aldehyde diethyl acetal
Assay ≥97.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC380930050 SDS Thermo Fisher Scientific
AC380930050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

87219-80-5
100.0
C10H20O2Si
200.35
3-Trimethylsilylpropargyl aldehyde diethyl acetal
97.0
97+%
(CH3)3SiCCCH(OCH2CH3)2
MFCD08275372
≥97.0%

Specifications

200.35
5g
97.0
Authentic
97+%
1.4300 to 1.4320 (20°C, 589nm)
(CH3)3SiCCCH(OCH2CH3)2
≥97.0%
Liquid
Clear colorless to slightly yellow
100.0
87219-80-5
C10H20O2Si
MFCD08275372
3-Trimethylsilylpropargyl aldehyde diethyl acetal

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning