Please login to your online account to display your discounted pricing

3-(Trimethylsilyloxy)-1-butyne 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(Trimethylsilyloxy)-1-butyne
CAS 17869-76-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7 H14 O Si
Formula Weight 142.27
MDL Number MFCD00190215
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC389240250

  SDS
thermo fisher scientific
AC389240250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 142.27
Quantity 25g
Color Clear pale yellow
Density 0.8180g/mL
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C7 H14 O Si
CAS 17869-76-0
Infrared Spectrum Authentic
Chemical Name or Material 3-(Trimethylsilyloxy)-1-butyne
Packaging Glass bottle
Refractive Index 1.4030 to 1.4050 (20°C, 589nm)
Assay Percent Range 97%
MDL Number MFCD00190215
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.