missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Trimethylsilyl)propargyl bromide, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 38002-45-8 |
---|---|
Molecular Formula | C6H11BrSi |
Molecular Weight (g/mol) | 191.14 |
MDL Number | MFCD00134460 |
InChI Key | GAPRPFRDVCCCHR-UHFFFAOYSA-N |
Synonym | 3-bromo-1-trimethylsilyl-1-propyne, 3-bromoprop-1-yn-1-yl trimethylsilane, 3-bromo-1-propynyl trimethylsilane, 3-trimethylsilyl propargyl bromide, 3-bromoprop-1-ynyl trimethyl silane, silane, 3-bromo-1-propynyl trimethyl, 3-bromoprop-1-yn-1-yl trimethyl silane, acmc-1cthe, ksc496i3r |
PubChem CID | 642589 |
IUPAC Name | 3-bromoprop-1-ynyl(trimethyl)silane |
SMILES | C[Si](C)(C)C#CCBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC399840010
|
Thermo Scientific Chemicals
399840010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC399840050
|
Thermo Scientific Chemicals
399840050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
38002-45-8 | |
191.14 | |
GAPRPFRDVCCCHR-UHFFFAOYSA-N | |
642589 | |
C[Si](C)(C)C#CCBr |
C6H11BrSi | |
MFCD00134460 | |
3-bromo-1-trimethylsilyl-1-propyne, 3-bromoprop-1-yn-1-yl trimethylsilane, 3-bromo-1-propynyl trimethylsilane, 3-trimethylsilyl propargyl bromide, 3-bromoprop-1-ynyl trimethyl silane, silane, 3-bromo-1-propynyl trimethyl, 3-bromoprop-1-yn-1-yl trimethyl silane, acmc-1cthe, ksc496i3r | |
3-bromoprop-1-ynyl(trimethyl)silane |
Specifications
38002-45-8 | |
44°C to 45°C (2.0mmHg) | |
96% min. (GC) | |
C6H11BrSi | |
MFCD00134460 | |
1.351 | |
Solubility in water: insoluble | |
C[Si](C)(C)C#CCBr | |
191.14 | |
191.14 | |
3-(Trimethylsilyl)propargyl bromide |
1.3510g/mL | |
62°C | |
Glass Bottle | |
(CH3)3SiC∽CCH2Br | |
1g | |
3-bromo-1-trimethylsilyl-1-propyne, 3-bromoprop-1-yn-1-yl trimethylsilane, 3-bromo-1-propynyl trimethylsilane, 3-trimethylsilyl propargyl bromide, 3-bromoprop-1-ynyl trimethyl silane, silane, 3-bromo-1-propynyl trimethyl, 3-bromoprop-1-yn-1-yl trimethyl silane, acmc-1cthe, ksc496i3r | |
GAPRPFRDVCCCHR-UHFFFAOYSA-N | |
3-bromoprop-1-ynyl(trimethyl)silane | |
642589 | |
97% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Warning
RUO â Research Use Only