missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trimethylsilyl)propargyl alcohol, 98+%

Manufacturer:  Alfa Aesar™ L0964914

 View more versions of this product

Catalog No. AAL0964914



Description

Description

Applications
3-(Trimethylsilyl)propargyl alcohol is used as a reagent to synthesize Isbogrel, a Thromboxane A 2 (TXA 2) synthase inhibitor and also a TXA 2 receptor antagonist that has potential to treat asthma.

Solubility
Not miscible with water.

Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Protect from heat. Store under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents, heat and moisture.
Specifications

Specifications

3-(Trimethylsilyl)propargyl alcohol
C6H12OSi
MFCD00002913
1902505
ZVGCJDPEKKEYES-UHFFFAOYSA-N
3-trimethylsilylprop-2-yn-1-ol
78930
≥98%
25g
5272-36-6
(CH3)3SiC≡CCH2OH
UN2810
3-trimethylsilyl-2-propyn-1-ol, 3-trimethylsilyl propargyl alcohol, 3-trimethylsilyl-2-propyn-1-ol, 2-propyn-1-ol, 3-trimethylsilyl, 3-trimethylsilyl prop-2-yn-1-ol, 1-trimethylsilylpropargyl alcohol, acmc-209l1b, 3-trimethylsilylpropargyl alcohol, 3-trimethylsilanyl-prop-2-yn-1-ol, 3-trimethylsilyl-2-propyn-1-ol #
C[Si](C)(C)C#CCO
128.246
128.25
170°C to 172°C
66°C (150°F)

Chemical Identifiers

5272-36-6
128.246
ZVGCJDPEKKEYES-UHFFFAOYSA-N
78930
C[Si](C)(C)C#CCO
C6H12OSi
MFCD00002913
3-trimethylsilyl-2-propyn-1-ol, 3-trimethylsilyl propargyl alcohol, 3-trimethylsilyl-2-propyn-1-ol, 2-propyn-1-ol, 3-trimethylsilyl, 3-trimethylsilyl prop-2-yn-1-ol, 1-trimethylsilylpropargyl alcohol, acmc-209l1b, 3-trimethylsilylpropargyl alcohol, 3-trimethylsilanyl-prop-2-yn-1-ol, 3-trimethylsilyl-2-propyn-1-ol #
3-trimethylsilylprop-2-yn-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280h-P305+P351+P338-P309-P310
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents