missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-[(Trimethylsilyl)ethynyl]pyridine, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5510114

 View more versions of this product

Catalog No. AAH5510114



Specifications

Specifications

3-[(Trimethylsilyl)ethynyl]pyridine
1.5290
Colorless to Yellow
80673-00-3
MFCD01863744
MTMBLPOPETZZRX-UHFFFAOYSA-N
trimethyl(2-pyridin-3-ylethynyl)silane
4100857
Liquid
0.922g/mL
229°C
25g
C10H13NSi
1-3-pyridyl-2-trimethylsilylacetylene
C[Si](C)(C)C#CC1=CN=CC=C1
175.306
175.31
97%

Chemical Identifiers

80673-00-3
175.306
MTMBLPOPETZZRX-UHFFFAOYSA-N
4100857
C[Si](C)(C)C#CC1=CN=CC=C1
C10H13NSi
MFCD01863744
1-3-pyridyl-2-trimethylsilylacetylene
trimethyl(2-pyridin-3-ylethynyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents