missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-[(Trimethylsilyl)ethynyl]benzonitrile, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H55012MD

 View more versions of this product

Catalog No. AAH55012MD



Specifications

Specifications

3-[(Trimethylsilyl)ethynyl]benzonitrile
250mg
C12H13NSi
3-2-trimethylsilyl ethynyl benzonitrile
C[Si](C)(C)C#CC1=CC(=CC=C1)C#N
199.328
199.33
42°C to 47°C
190771-22-3
MFCD09260494
QEZDMZVLNWSJSA-UHFFFAOYSA-N
3-(2-trimethylsilylethynyl)benzonitrile
22570318
97%

Chemical Identifiers

190771-22-3
199.328
QEZDMZVLNWSJSA-UHFFFAOYSA-N
22570318
C[Si](C)(C)C#CC1=CC(=CC=C1)C#N
C12H13NSi
MFCD09260494
3-2-trimethylsilyl ethynyl benzonitrile
3-(2-trimethylsilylethynyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents