missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-[(Trimethylsilyl)ethynyl]benzonitrile, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5501206

 View more versions of this product

Catalog No. AAH5501206



Specifications

Specifications

3-[(Trimethylsilyl)ethynyl]benzonitrile
5g
C12H13NSi
3-2-trimethylsilyl ethynyl benzonitrile
C[Si](C)(C)C#CC1=CC(=CC=C1)C#N
199.328
199.33
42°C to 47°C
190771-22-3
MFCD09260494
QEZDMZVLNWSJSA-UHFFFAOYSA-N
3-(2-trimethylsilylethynyl)benzonitrile
22570318
97%

Chemical Identifiers

190771-22-3
199.328
QEZDMZVLNWSJSA-UHFFFAOYSA-N
22570318
C[Si](C)(C)C#CC1=CC(=CC=C1)C#N
C12H13NSi
MFCD09260494
3-2-trimethylsilyl ethynyl benzonitrile
3-(2-trimethylsilylethynyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents