Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Trimethylsilyl-2-oxazolidinone

$63.59 - $63.59

Chemical Identifiers

Chemical Name or Material 3-Trimethylsilyl-2-oxazolidinone
CAS 43112-38-5
Formula Weight 159.26
MDL Number MFCD00010197
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4120906

  SDS
alfa aesar™
4120906
5g Each for $63.59
Description & Specifications

Specifications

Formula Weight 159.26
Refractive Index 1.4540
Density 1.013g/mL
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Flash Point 92° (198°F)
Sensitivity Moisture Sensitive
Quantity 5g
Physical Form Liquid
CAS 43112-38-5
Health Hazard 1 Warning
Chemical Name or Material 3-Trimethylsilyl-2-oxazolidinone
MDL Number MFCD00010197
Boiling Point 99° to 100°C (6mmHg)
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.