missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Trimethylsilyl)-2-naphthyl Trifluoromethanesulfonate 95.0+%, TCI America™
$481.11 - $1668.95
Chemical Identifiers
CAS | 780820-43-1 |
---|---|
Molecular Formula | C14H15F3O3SSi |
Molecular Weight (g/mol) | 348.411 |
InChI Key | OSZYFPWWHQEAGW-UHFFFAOYSA-N |
Synonym | 3-(Trimethylsilyl)-2-naphthyl Triflate, Trifluoromethanesulfonic Acid 3-(Trimethylsilyl)-2-naphthyl Ester |
PubChem CID | 44629848 |
IUPAC Name | (3-trimethylsilylnaphthalen-2-yl) trifluoromethanesulfonate |
SMILES | C[Si](C)(C)C1=CC2=CC=CC=C2C=C1OS(=O)(=O)C(F)(F)F |
Chemical Identifiers
780820-43-1 | |
348.411 | |
3-(Trimethylsilyl)-2-naphthyl Triflate, Trifluoromethanesulfonic Acid 3-(Trimethylsilyl)-2-naphthyl Ester | |
(3-trimethylsilylnaphthalen-2-yl) trifluoromethanesulfonate |
C14H15F3O3SSi | |
OSZYFPWWHQEAGW-UHFFFAOYSA-N | |
44629848 | |
C[Si](C)(C)C1=CC2=CC=CC=C2C=C1OS(=O)(=O)C(F)(F)F |
Specifications
780820-43-1 | |
C14H15F3O3SSi | |
3265 | |
OSZYFPWWHQEAGW-UHFFFAOYSA-N | |
(3-trimethylsilylnaphthalen-2-yl) trifluoromethanesulfonate | |
44629848 | |
≥95.0% (GC) | |
3-(Trimethylsilyl)-2-naphthyl Trifluoromethanesulfonate |
Yellow | |
1 g | |
3-(Trimethylsilyl)-2-naphthyl Triflate, Trifluoromethanesulfonic Acid 3-(Trimethylsilyl)-2-naphthyl Ester | |
C[Si](C)(C)C1=CC2=CC=CC=C2C=C1OS(=O)(=O)C(F)(F)F | |
348.411 | |
348.41 | |
Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator