Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Trimethylsilyl-1-propylboronic acid pinacol ester, 94%

$163.16 - $163.16

Chemical Identifiers

Chemical Name or Material 3-Trimethylsilyl-1-propylboronic acid pinacol ester
Molecular Formula C12H27BO2Si
Formula Weight 242.24
MDL Number MFCD09953502
Synonym 4,4,5,5-Tetramethyl-2-(3-trimethylsilylpropyl)-1,3,2-dioxoborolane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2841306

 
alfa aesar™
H2841306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH28413MD

 
alfa aesar™
H28413MD
250mg Each for $163.16

AAH2841303

 
alfa aesar™
H2841303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 242.24
Chemical Name or Material 3-Trimethylsilyl-1-propylboronic acid pinacol ester
Assay 94%
Molecular Formula C12H27BO2Si
MDL Number MFCD09953502
Synonym 4,4,5,5-Tetramethyl-2-(3-trimethylsilylpropyl)-1,3,2-dioxoborolane
Quantity 5g