Please login to your online account to display your discounted pricing

3-(Trimethoxysilyl)propyl acrylate, 90%, stabilized, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC426600250

 View more versions of this product

Catalog No. AC426600250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥90%
Formula Weight 234.32
Quantity 25g
Color Clear colorless
Density 1.0590g/mL
CAS Min % 88.0
CAS Max % 100.0
Chemical Name or Material 3-(Trimethoxysilyl)propyl acrylate
Packaging Glass bottle
Refractive Index 1.4280 to 1.4300 (20°C, 589nm)
Name Note Stabilized
Assay Percent Range 90%
Molecular Formula C9H18O5Si
Linear Formula H2C=CHCO2(CH2)3Si(OCH3)3
MDL Number MFCD00054803
CAS 4369-14-6
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Stabilizer stabilized with BHT

Specifications

Chemical Name or Material 3-(Trimethoxysilyl)propyl acrylate
Name Note Stabilized
CAS 4369-14-6
CAS Min % 88.0
CAS Max % 100.0
Assay ≥90%
Assay Percent Range 90%
Molecular Formula C9H18O5Si
Linear Formula H2C=CHCO2(CH2)3Si(OCH3)3
Formula Weight 234.32
MDL Number MFCD00054803
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.

SDS & Safety Documents