Learn More
3-(Trifluoromethyl)thiobenzamide, 97%, Thermo Scientific Chemicals
$55.97 - $327.30
Chemical Identifiers
CAS | 53515-17-6 |
---|---|
Molecular Formula | C8H6F3NS |
Molecular Weight (g/mol) | 205.20 |
MDL Number | MFCD01567258 |
InChI Key | WOJQBHSZMFRQIR-UHFFFAOYSA-N |
Synonym | 3-trifluoromethyl thiobenzamide, 3-trifluoromethylthiobenzamide, 3-trifluoromethyl benzenecarbothioamide, 3-trifluoromethyl benzene-1-carbothioamide, 3-trifluoromethyl-thiobenzamide, amino 3-trifluoromethyl phenyl methane-1-thione, acmc-20amye, pubchem5483, maybridge1_008312 |
PubChem CID | 2777849 |
IUPAC Name | 3-(trifluoromethyl)benzenecarbothioamide |
SMILES | NC(=S)C1=CC(=CC=C1)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH26311MD
|
Thermo Scientific Chemicals
H26311MD |
250 mg |
N/A
|
|
|||||
AAH2631103
|
Thermo Scientific Chemicals
H2631103 |
1 g |
N/A
|
|
|||||
AAH2631106
|
Thermo Scientific Chemicals
H2631106 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilityInsoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
Chemical Identifiers
53515-17-6 | |
205.20 | |
WOJQBHSZMFRQIR-UHFFFAOYSA-N | |
2777849 | |
NC(=S)C1=CC(=CC=C1)C(F)(F)F |
C8H6F3NS | |
MFCD01567258 | |
3-trifluoromethyl thiobenzamide, 3-trifluoromethylthiobenzamide, 3-trifluoromethyl benzenecarbothioamide, 3-trifluoromethyl benzene-1-carbothioamide, 3-trifluoromethyl-thiobenzamide, amino 3-trifluoromethyl phenyl methane-1-thione, acmc-20amye, pubchem5483, maybridge1_008312 | |
3-(trifluoromethyl)benzenecarbothioamide |
Specifications
53515-17-6 | |
MFCD01567258 | |
UN2811 | |
Insoluble in water. | |
NC(=S)C1=CC(=CC=C1)C(F)(F)F | |
205.20 | |
205.2 | |
3-(Trifluoromethyl)thiobenzamide |
C8H6F3NS | |
250 mg | |
3-trifluoromethyl thiobenzamide, 3-trifluoromethylthiobenzamide, 3-trifluoromethyl benzenecarbothioamide, 3-trifluoromethyl benzene-1-carbothioamide, 3-trifluoromethyl-thiobenzamide, amino 3-trifluoromethyl phenyl methane-1-thione, acmc-20amye, pubchem5483, maybridge1_008312 | |
WOJQBHSZMFRQIR-UHFFFAOYSA-N | |
3-(trifluoromethyl)benzenecarbothioamide | |
2777849 | |
97% |
Safety and Handling
GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only