missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethyl)pyridine, 97%

Manufacturer:  Alfa Aesar™ L1728006

 View more versions of this product

Catalog No. AAL1728006



Specifications

Specifications

3-(Trifluoromethyl)pyridine
111°C to 114°C
3796-23-4
MFCD00100032
1563102
JTZSFNHHVULOGJ-UHFFFAOYSA-N
3-(trifluoromethyl)pyridine
77417
97%
19°C (66°F)
5g
C6H4F3N
UN1993
3-cf3-pyridine
C1=CC(=CN=C1)C(F)(F)F
147.1
147.1

Chemical Identifiers

3796-23-4
147.1
JTZSFNHHVULOGJ-UHFFFAOYSA-N
77417
C1=CC(=CN=C1)C(F)(F)F
C6H4F3N
MFCD00100032
3-cf3-pyridine
3-(trifluoromethyl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Documents