missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethyl)pyridine-4-carboxylic acid, 95%

$52.12 - $616.85

Chemical Identifiers

CAS 590371-38-3
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.109
MDL Number MFCD08234940
InChI Key FBOSFEOQEZQFOC-UHFFFAOYSA-N
Synonym 3-trifluoromethyl pyridine-4-carboxylic acid, 3-trifluoromethyl isonicotinic acid, 3-trifluoromethyl-isonicotinic acid, 3-trifluoromethyl-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 3-trifluoromethyl, pubchem22216, ksc268s9d, 3-trifluoromethylisonicotinic acid, 3-trifluoromethyl isonicotinicacid, 3-trifluoromethyl isonicotinic acid
PubChem CID 25210500
IUPAC Name 3-(trifluoromethyl)pyridine-4-carboxylic acid
SMILES C1=CN=CC(=C1C(=O)O)C(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63174MD View Documents Alfa Aesar™
H63174MD
250mg Each for $52.12
 
AAH6317403 View Documents Alfa Aesar™
H6317403
1g Each for $154.22
 
AAH6317406 View Documents Alfa Aesar™
H6317406
5g Each for $616.85
 
Specifications

Chemical Identifiers

590371-38-3
191.109
FBOSFEOQEZQFOC-UHFFFAOYSA-N
25210500
C1=CN=CC(=C1C(=O)O)C(F)(F)F
C7H4F3NO2
MFCD08234940
3-trifluoromethyl pyridine-4-carboxylic acid, 3-trifluoromethyl isonicotinic acid, 3-trifluoromethyl-isonicotinic acid, 3-trifluoromethyl-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 3-trifluoromethyl, pubchem22216, ksc268s9d, 3-trifluoromethylisonicotinic acid, 3-trifluoromethyl isonicotinicacid, 3-trifluoromethyl isonicotinic acid
3-(trifluoromethyl)pyridine-4-carboxylic acid

Specifications

250mg
C7H4F3NO2
3-trifluoromethyl pyridine-4-carboxylic acid, 3-trifluoromethyl isonicotinic acid, 3-trifluoromethyl-isonicotinic acid, 3-trifluoromethyl-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 3-trifluoromethyl, pubchem22216, ksc268s9d, 3-trifluoromethylisonicotinic acid, 3-trifluoromethyl isonicotinicacid, 3-trifluoromethyl isonicotinic acid
C1=CN=CC(=C1C(=O)O)C(F)(F)F
191.109
191.11
3-(Trifluoromethyl)pyridine-4-carboxylic acid
590371-38-3
MFCD08234940
FBOSFEOQEZQFOC-UHFFFAOYSA-N
3-(trifluoromethyl)pyridine-4-carboxylic acid
25210500
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning