missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trifluoromethyl)pyridine-2-carboxylic Acid, TCI America™

Manufacturer:  TCI America T3155500MG

 View more versions of this product

Catalog No. T3155500MG



Specifications

Specifications

3-(Trifluoromethyl)pyridine-2-carboxylic Acid
113°C
87407-12-3
MFCD07375386
DMWLBOPLZYJGPT-UHFFFAOYSA-N
3-(trifluoromethyl)pyridine-2-carboxylic acid
13233844
Crystal-Powder at 20°C
White-Yellow
500mg
C7H4F3NO2
3-trifluoromethyl pyridine-2-carboxylic acid, 3-trifluoromethyl picolinic acid, 3-trifluoromethyl-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 3-trifluoromethyl, 3-trifluoromethyl-2-picolinic acid, 3-trifluoromethyl pyridine-2-carboxylic acid, acmc-209qkb, ksc447o0j, 3-trifluoromethylpicolinic acid, trifluoromethyl-2-picolinic acid
C1=CC(=C(N=C1)C(=O)O)C(F)(F)F
191.109
191.11
≥98.0% (GC,T)

Chemical Identifiers

87407-12-3
191.109
DMWLBOPLZYJGPT-UHFFFAOYSA-N
13233844
C1=CC(=C(N=C1)C(=O)O)C(F)(F)F
C7H4F3NO2
MFCD07375386
3-trifluoromethyl pyridine-2-carboxylic acid, 3-trifluoromethyl picolinic acid, 3-trifluoromethyl-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 3-trifluoromethyl, 3-trifluoromethyl-2-picolinic acid, 3-trifluoromethyl pyridine-2-carboxylic acid, acmc-209qkb, ksc447o0j, 3-trifluoromethylpicolinic acid, trifluoromethyl-2-picolinic acid
3-(trifluoromethyl)pyridine-2-carboxylic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents