Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-(Trifluoromethyl)propiophenone, 97%

$91.73 - $338.64

Chemical Identifiers

Chemical Name or Material 3'-(Trifluoromethyl)propiophenone
CAS 1533-03-5
Molecular Formula C10H9F3O
Formula Weight 202.18
MDL Number MFCD00039227
Beilstein 2101539
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2014706

  SDS
alfa aesar™
B2014706
5g Each for $91.73

AAB2014714

  SDS
alfa aesar™
B2014714
25g Each for $338.64
Description & Specifications

Specifications

Formula Weight 202.18
Chemical Name or Material 3'-(Trifluoromethyl)propiophenone
Assay 97%
Molecular Formula C10H9F3O
MDL Number MFCD00039227
CAS 1533-03-5
Beilstein 2101539
Flash Point 108°C (226°F)
Boiling Point 216° to 217°C
Melting Point 18° to 19°C
Quantity 5g