missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ [3-(Trifluoromethyl)phenylthio]acetic acid

$134.38 - $606.47

Chemical Identifiers

CAS 349-83-7
Molecular Formula C9H7F3O2S
Formula Weight 236.21
MDL Number MFCD00052114
Chemical Name or Material [3-(Trifluoromethyl)phenylthio]acetic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5113503 SDS Alfa Aesar™
H5113503
1g Each for $134.38
AAH5113506 SDS Alfa Aesar™
H5113506
5g Each for $606.47
Specifications Description & Specifications

Chemical Identifiers

349-83-7
236.21
[3-(Trifluoromethyl)phenylthio]acetic acid
C9H7F3O2S
MFCD00052114

Specifications

1g
C9H7F3O2S
MFCD00052114
349-83-7
236.21
[3-(Trifluoromethyl)phenylthio]acetic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning