Please login to your online account to display your discounted pricing

3-(Trifluoromethyl)phenylhydrazine, 90%, Tech., ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC333100050

 View more versions of this product

Catalog No. AC333100050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Physical Form Liquid
Assay ≥88%
Formula Weight 176.14
Quantity 5g
Color Clear yellow to orange to brown
Density 1.348g/mL
CAS Min % 88.0
CAS Max % 100.0
Chemical Name or Material 3-(Trifluoromethyl)phenylhydrazine
Assay Percent Range 90%
Linear Formula CF3C6H4NHNH
Molecular Formula C7H7F3N2
MDL Number MFCD00025093
CAS 368-78-5
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.

Specifications

Chemical Name or Material 3-(Trifluoromethyl)phenylhydrazine
CAS 368-78-5
CAS Min % 88.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C7H7F3N2
Linear Formula CF3C6H4NHNH
Formula Weight 176.14
MDL Number MFCD00025093
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.

SDS & Safety Documents