Please login to your online account to display your discounted pricing

3-(Trifluoromethyl)phenylhydrazine, 90%, Tech., ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethyl)phenylhydrazine
CAS 368-78-5
CAS Min % 88.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C7H7F3N2
Linear Formula CF3C6H4NHNH
Formula Weight 176.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC333100010

  SDS
thermo fisher scientific
333100010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥88%
Purity Grade Technical Grade
Physical Form Liquid
Formula Weight 176.14
Quantity 1g
Color Clear yellow to orange to brown
Density 1.348g/mL
CAS Min % 88.0
CAS Max % 100.0
Chemical Name or Material 3-(Trifluoromethyl)phenylhydrazine
Assay Percent Range 90%
Molecular Formula C7H7F3N2
Linear Formula CF3C6H4NHNH
CAS 368-78-5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00025093
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.