missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trifluoromethyl)phenylacetonitrile, TCI America™

Manufacturer:  TCI America T236725G

 View more versions of this product

Catalog No. T236725G



Specifications

Specifications

3-(Trifluoromethyl)phenylacetonitrile
Yellow
2338-76-3
MFCD00001913
3-trifluoromethyl phenylacetonitrile, 3-trifluoromethylbenzylcyanide, 2-3-trifluoromethyl phenyl acetonitrile, benzeneacetonitrile, 3-trifluoromethyl, 3-trifluoromethyl benzyl cyanide, m-trifluoromethylphenylacetonitrile, m-trifluoromethylbenzyl cyanide, 3-trifluoromethylphenylacetonitrile, 3-trifluormethylbenzylcyanide, 3-trifluoromethyl benzeneacetonitrile
C1=CC(=CC(=C1)C(F)(F)F)CC#N
185.149
185.15
≥98.0% (GC)
93°C
25g
C9H6F3N
1992
JOIYKSLWXLFGGR-UHFFFAOYSA-N
2-[3-(trifluoromethyl)phenyl]acetonitrile
75360
Clear Liquid at 20°C

Chemical Identifiers

2338-76-3
185.149
JOIYKSLWXLFGGR-UHFFFAOYSA-N
75360
C1=CC(=CC(=C1)C(F)(F)F)CC#N
C9H6F3N
MFCD00001913
3-trifluoromethyl phenylacetonitrile, 3-trifluoromethylbenzylcyanide, 2-3-trifluoromethyl phenyl acetonitrile, benzeneacetonitrile, 3-trifluoromethyl, 3-trifluoromethyl benzyl cyanide, m-trifluoromethylphenylacetonitrile, m-trifluoromethylbenzyl cyanide, 3-trifluoromethylphenylacetonitrile, 3-trifluormethylbenzylcyanide, 3-trifluoromethyl benzeneacetonitrile
2-[3-(trifluoromethyl)phenyl]acetonitrile
Safety and Handling

Safety and Handling

RTECSNumber : CY1820000

TSCA : Yes

Documents