missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethyl)phenylacetonitrile, 97%

Manufacturer:  Alfa Aesar™ A1466318

 View more versions of this product

Catalog No. AAA1466318



Specifications

Specifications

3-(Trifluoromethyl)phenylacetonitrile
92°C to 93°C (4mmHg)
2338-76-3
MFCD00001913
909640
JOIYKSLWXLFGGR-UHFFFAOYSA-N
2-[3-(trifluoromethyl)phenyl]acetonitrile
75360
97%
>100°C (212°F)
50g
C9H6F3N
UN3276
3-trifluoromethyl phenylacetonitrile, 3-trifluoromethylbenzylcyanide, 2-3-trifluoromethyl phenyl acetonitrile, benzeneacetonitrile, 3-trifluoromethyl, 3-trifluoromethyl benzyl cyanide, m-trifluoromethylphenylacetonitrile, m-trifluoromethylbenzyl cyanide, 3-trifluoromethylphenylacetonitrile, 3-trifluormethylbenzylcyanide, 3-trifluoromethyl benzeneacetonitrile
C1=CC(=CC(=C1)C(F)(F)F)CC#N
185.149
185.15

Chemical Identifiers

2338-76-3
185.149
JOIYKSLWXLFGGR-UHFFFAOYSA-N
75360
C1=CC(=CC(=C1)C(F)(F)F)CC#N
C9H6F3N
MFCD00001913
3-trifluoromethyl phenylacetonitrile, 3-trifluoromethylbenzylcyanide, 2-3-trifluoromethyl phenyl acetonitrile, benzeneacetonitrile, 3-trifluoromethyl, 3-trifluoromethyl benzyl cyanide, m-trifluoromethylphenylacetonitrile, m-trifluoromethylbenzyl cyanide, 3-trifluoromethylphenylacetonitrile, 3-trifluormethylbenzylcyanide, 3-trifluoromethyl benzeneacetonitrile
2-[3-(trifluoromethyl)phenyl]acetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

RTECSNumber : CY1820000

TSCA : TSCA

Documents