Learn More
3-(Trifluoromethyl)phenylacetonitrile, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1466314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-(Trifluoromethyl)phenylacetonitrile | |
1.187 | |
>100°C (212°F) | |
1.459 | |
25 g | |
909640 | |
JOIYKSLWXLFGGR-UHFFFAOYSA-N | |
2-[3-(trifluoromethyl)phenyl]acetonitrile | |
75360 | |
97% |
2338-76-3 | |
92°C to 93°C (4 mmHg) | |
C9H6F3N | |
MFCD00001913 | |
UN3276 | |
3-trifluoromethyl phenylacetonitrile, 3-trifluoromethylbenzylcyanide, 2-3-trifluoromethyl phenyl acetonitrile, benzeneacetonitrile, 3-trifluoromethyl, 3-trifluoromethyl benzyl cyanide, m-trifluoromethylphenylacetonitrile, m-trifluoromethylbenzyl cyanide, 3-trifluoromethylphenylacetonitrile, 3-trifluormethylbenzylcyanide, 3-trifluoromethyl benzeneacetonitrile | |
FC(F)(F)C1=CC=CC(CC#N)=C1 | |
185.15 | |
185.15 |
Chemical Identifiers
2338-76-3 | |
185.15 | |
JOIYKSLWXLFGGR-UHFFFAOYSA-N | |
75360 | |
FC(F)(F)C1=CC=CC(CC#N)=C1 |
C9H6F3N | |
MFCD00001913 | |
3-trifluoromethyl phenylacetonitrile, 3-trifluoromethylbenzylcyanide, 2-3-trifluoromethyl phenyl acetonitrile, benzeneacetonitrile, 3-trifluoromethyl, 3-trifluoromethyl benzyl cyanide, m-trifluoromethylphenylacetonitrile, m-trifluoromethylbenzyl cyanide, 3-trifluoromethylphenylacetonitrile, 3-trifluormethylbenzylcyanide, 3-trifluoromethyl benzeneacetonitrile | |
2-[3-(trifluoromethyl)phenyl]acetonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 219-053-8
RTECSNumber : CY1820000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only