Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethyl)phenylacetone, 97%, may cont. up to ca 5% monohydrate

$106.08 - $370.66

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethyl)phenylacetone
CAS 21906-39-8
Molecular Formula C10H9F3O
Formula Weight 202.18
MDL Number MFCD00000397
Synonym 1-(alpha,alpha,alpha-Trifluoro-m-tolyl)-2-propanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2709514

  SDS
alfa aesar™
H2709514
25g Each for $370.66

AAH2709506

  SDS
alfa aesar™
H2709506
5g Each for $106.08
Description & Specifications

Specifications

Formula Weight 202.18
Chemical Name or Material 3-(Trifluoromethyl)phenylacetone
Assay 97%
Assay Percent Notes may contain up to ≈5% monohydrate
Molecular Formula C10H9F3O
MDL Number MFCD00000397
CAS 21906-39-8
Synonym 1-(alpha,alpha,alpha-Trifluoro-m-tolyl)-2-propanone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)
Boiling Point 89° to 90°C (0.5mmHg)
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.