Please login to your online account to display your discounted pricing

3-(Trifluoromethyl)phenylacetone, 97%, Acros Organics

$227.15 - $227.15

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethyl)phenylacetone
CAS 21906-39-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H9F3O
Linear Formula CF3C6H4CH2COCH3
Formula Weight 202.18
MDL Number MFCD00000397
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC186160025

  SDS
thermo fisher scientific
AC186160025
Glass bottle 2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC186160010

  SDS
thermo fisher scientific
AC186160010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC186160100

  SDS
thermo fisher scientific
186160100
Glass bottle 10g Each for $227.15
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 202.18
Quantity 2.5g
Color Clear yellow
Density 1.204g/mL
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C10H9F3O
Synonym 1-(α, α, α-Trifluoro-m-tolyl)-2-propanone
Chemical Name or Material 3-(Trifluoromethyl)phenylacetone
Assay Percent Range 97%
CAS 21906-39-8
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula CF3C6H4CH2COCH3
MDL Number MFCD00000397
Packaging Glass bottle
Refractive Index 1.4560 to 1.4580 (20°C, 589nm)
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.