missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trifluoromethyl)phenyl isothiocyanate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1164603

 View more versions of this product

Catalog No. AAL1164603



Description

Description

Used as pharamaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharamaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
Specifications

Specifications

3-(Trifluoromethyl)phenyl isothiocyanate
1.335
95°C (203°F)
C8H4F3NS
MFCD00004806
UN2922
Moisture sensitive
Hydrolyzes in water.
C1=CC(=CC(=C1)N=C=S)C(F)(F)F
203.182
203.19
1840-19-3
208°C
Pungent
1.5547
1 g
781205
3-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl benzene, 3-trifluoromethyl phenylisothiocyanate, benzene, 1-isothiocyanato-3-trifluoromethyl, m-trifluoromethyl phenyl isothiocyanate, alpha,alpha,alpha-trifluoro-m-tolyl isothiocyanate, 3-trifluoromethylphenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl-benzene, benzene,1-isothiocyanato-3-trifluoromethyl
GFEPANUKFYVALF-UHFFFAOYSA-N
1-isothiocyanato-3-(trifluoromethyl)benzene
137232
98%

Chemical Identifiers

1840-19-3
203.182
GFEPANUKFYVALF-UHFFFAOYSA-N
137232
C1=CC(=CC(=C1)N=C=S)C(F)(F)F
C8H4F3NS
MFCD00004806
3-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl benzene, 3-trifluoromethyl phenylisothiocyanate, benzene, 1-isothiocyanato-3-trifluoromethyl, m-trifluoromethyl phenyl isothiocyanate, alpha,alpha,alpha-trifluoro-m-tolyl isothiocyanate, 3-trifluoromethylphenyl isothiocyanate, 1-isothiocyanato-3-trifluoromethyl-benzene, benzene,1-isothiocyanato-3-trifluoromethyl
1-isothiocyanato-3-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only