Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethyl)phenyl isocyanate, 98%

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethyl)phenyl isocyanate
CAS 329-01-1
Molecular Formula C8H4F3NO
Formula Weight 187.12
MDL Number MFCD00002020
UN Number UN2285
Beilstein 744880
Synonym 3-Isocyanatobenzotrifluoride; alpha,alpha,alpha-Trifluoro-m-tolyl isocyanate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0944803

 
alfa aesar™
L0944803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0944806

 
alfa aesar™
L0944806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0944814

 
alfa aesar™
L0944814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 187.12
TSCA TSCA
Assay 98%
Molecular Formula C8H4F3NO
CAS 329-01-1
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H330-H334-H226-H315-H319-H335
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Odor Pungent
MDL Number MFCD00002020
Beilstein 744880
Synonym 3-Isocyanatobenzotrifluoride; alpha,alpha,alpha-Trifluoro-m-tolyl isocyanate
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATOBENZOTRIFLUORIDES
RTECS Number NR0200000
Flash Point 58°C (136°F)
Boiling Point 53° to 54°C (11mmHg)
Quantity 1g
Chemical Name or Material 3-(Trifluoromethyl)phenyl isocyanate
UN Number UN2285
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H330-H334-H226-H315-H319-H335
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C