Please login to your online account to display your discounted pricing

3-(Trifluoromethyl)phenethylamine, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC365520050

 View more versions of this product

Catalog No. AC365520050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 189.18
Quantity 5g
Color Clear colorless to pale yellow
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
Refractive Index 1.4635 to 1.4655 (20°C, 589nm)
Assay Percent Range 98%
Chemical Name or Material 3-(Trifluoromethyl)phenethylamine
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
MDL Number MFCD00040756
CAS 52516-30-0
Molecular Formula C9H10F3N

Specifications

Chemical Name or Material 3-(Trifluoromethyl)phenethylamine
Name Note 98%
CAS 52516-30-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H10F3N
Formula Weight 189.18
MDL Number MFCD00040756
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents