missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trifluoromethyl)phenethylamine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC365520010

 View more versions of this product

Catalog No. AC365520010



Description

Description

Specifications

Specifications

3-(Trifluoromethyl)phenethylamine
Glass bottle
Colorless to Yellow
98%
97.5
97.5% min. (GC)
MFCD00040756
BPVYCXMGJPKOTQ-UHFFFAOYSA-N
2-[3-(trifluoromethyl)phenyl]ethanamine
104223
Liquid
Authentic
1.4635 to 1.4655
1g
52516-30-0
100.0
C9H10F3N
2-3-trifluoromethyl phenyl ethanamine, 2-3-trifluoromethyl phenyl ethylamine, 2-3-trifluoromethylphenyl ethylamine, 2-3-trifluoromethylphenyl-ethylamine, 3-trifluoromethyl phenethylamine, benzeneethanamine, 3-trifluoromethyl, m-trifluoromethyl phenethylamine, m-trifluoromethylphenethylamine, phenethylamine, m-trifluoromethyl
C1=CC(=CC(=C1)C(F)(F)F)CCN
189.18
189.18
98%

Chemical Identifiers

52516-30-0
189.18
BPVYCXMGJPKOTQ-UHFFFAOYSA-N
104223
C1=CC(=CC(=C1)C(F)(F)F)CCN
C9H10F3N
MFCD00040756
2-3-trifluoromethyl phenyl ethanamine, 2-3-trifluoromethyl phenyl ethylamine, 2-3-trifluoromethylphenyl ethylamine, 2-3-trifluoromethylphenyl-ethylamine, 3-trifluoromethyl phenethylamine, benzeneethanamine, 3-trifluoromethyl, m-trifluoromethyl phenethylamine, m-trifluoromethylphenethylamine, phenethylamine, m-trifluoromethyl
2-[3-(trifluoromethyl)phenyl]ethanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 257-988-3

SDS Documents