Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethyl)-D-phenylalanine, 97%

$213.96 - $213.96

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethyl)-D-phenylalanine
CAS 14464-67-6
Molecular Formula C10H10F3NO2
Formula Weight 233.19
MDL Number MFCD01860875
Synonym (R)-2-Amino-3-[3-(trifluoromethyl)phenyl]propionic acid; H-D-Phe(3-CF3)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2750803

  SDS
alfa aesar™
H2750803
1g Each for $213.96
Description & Specifications

Specifications

Formula Weight 233.19
Chemical Name or Material 3-(Trifluoromethyl)-D-phenylalanine
Assay 97%
Molecular Formula C10H10F3NO2
MDL Number MFCD01860875
CAS 14464-67-6
Synonym (R)-2-Amino-3-[3-(trifluoromethyl)phenyl]propionic acid; H-D-Phe(3-CF3)-OH
Quantity 1g