missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trifluoromethyl)cinnamic Acid, TCI America™

Manufacturer:  TCI America T154725G

 View more versions of this product

Catalog No. T154725G



Specifications

Specifications

3-(Trifluoromethyl)cinnamic Acid
135°C
779-89-5
MFCD00004393
KSBWHDDGWSYETA-SNAWJCMRSA-N
(E)-3-[3-(trifluoromethyl)phenyl]prop-2-enoic acid
719451
216.16
≥99.0% (GC,T)
White
25g
C10H7F3O2
3-trifluoromethyl cinnamic acid, 3-3-trifluoromethyl phenyl acrylic acid, m-trifluoromethyl cinnamic acid, e-3-trifluoromethyl cinnamic acid, unii-p9lj9on37w, 2e-3-3-trifluoromethyl phenyl prop-2-enoic acid, e-m-trifluoromethyl cinnamic acid, 3-e-trifluoromethyl cinnamic acid, 3-trifluoromethyl cinnamic acid, 3-3-trifluoromethyl phenyl acrylic acid
C1=CC(=CC(=C1)C(F)(F)F)C=CC(=O)O
216.159
CHEBI:60704
Crystal-Powder at 20°C

Chemical Identifiers

779-89-5
216.159
KSBWHDDGWSYETA-SNAWJCMRSA-N
719451
(E)-3-[3-(trifluoromethyl)phenyl]prop-2-enoic acid
C10H7F3O2
MFCD00004393
3-trifluoromethyl cinnamic acid, 3-3-trifluoromethyl phenyl acrylic acid, m-trifluoromethyl cinnamic acid, e-3-trifluoromethyl cinnamic acid, unii-p9lj9on37w, 2e-3-3-trifluoromethyl phenyl prop-2-enoic acid, e-m-trifluoromethyl cinnamic acid, 3-e-trifluoromethyl cinnamic acid, 3-trifluoromethyl cinnamic acid, 3-3-trifluoromethyl phenyl acrylic acid
CHEBI:60704
C1=CC(=CC(=C1)C(F)(F)F)C=CC(=O)O
Safety and Handling

Safety and Handling

TSCA : Yes

Documents