Please login to your online account to display your discounted pricing

3'-(Trifluoromethyl)biphenyl-4-sulfonyl chloride, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 3'-(Trifluoromethyl)biphenyl-4-sulfonyl chloride
Assay Percent Range 97%
Molecular Formula C13H8ClF3O2S
Formula Weight 320.72
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC450510010

  SDS
thermo fisher scientific
AC450510010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC450510050

  SDS
thermo fisher scientific
AC450510050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 320.72
Quantity 1g
Assay Percent Range 97%
Chemical Name or Material 3'-(Trifluoromethyl)biphenyl-4-sulfonyl chloride
Molecular Formula C13H8ClF3O2S
CAS 885267-96-9