missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3'-(Trifluoromethyl)biphenyl-4-sulfonyl chloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC450510050

 View more versions of this product

Catalog No. AC450510050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3'-(Trifluoromethyl)biphenyl-4-sulfonyl chloride
5g
96% min. diethylsulfonamide derivative (HPLC)
1,1'-biphenyl-4-sulfonylchloride, 3'-trifluoromethyl, 3'-trifluoromethyl 1,1'-biphenyl-4-sulfonyl chloride, 3'-trifluoromethyl biphenyl-4-sulfonyl chloride, 3'-trifluoromethyl-1,1'-biphenyl-4-sulfonyl chloride, 3'-trifluoromethylbiphenyl-4-sulfonyl chloride, 4-3-trifluoromethyl phenyl benzenesulfonyl chloride
C1=CC(=CC(=C1)C(F)(F)F)C2=CC=C(C=C2)S(=O)(=O)Cl
320.72
320.72
78°C to 82°C
885267-96-9
C13H8ClF3O2S
WJSSWLPZDVPLIK-UHFFFAOYSA-N
4-[3-(trifluoromethyl)phenyl]benzenesulfonyl chloride
16244690
97%

Chemical Identifiers

885267-96-9
320.72
1,1'-biphenyl-4-sulfonylchloride, 3'-trifluoromethyl, 3'-trifluoromethyl 1,1'-biphenyl-4-sulfonyl chloride, 3'-trifluoromethyl biphenyl-4-sulfonyl chloride, 3'-trifluoromethyl-1,1'-biphenyl-4-sulfonyl chloride, 3'-trifluoromethylbiphenyl-4-sulfonyl chloride, 4-3-trifluoromethyl phenyl benzenesulfonyl chloride
4-[3-(trifluoromethyl)phenyl]benzenesulfonyl chloride
C13H8ClF3O2S
WJSSWLPZDVPLIK-UHFFFAOYSA-N
16244690
C1=CC(=CC(=C1)C(F)(F)F)C2=CC=C(C=C2)S(=O)(=O)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

GHS Signal Word: Danger

SDS Documents