Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethyl)benzoylacetonitrile, 97%

$84.82 - $1,506.76

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethyl)benzoylacetonitrile
CAS 27328-86-5
Molecular Formula C10H6F3NO
Formula Weight 213.16
MDL Number MFCD00052315
Beilstein 2373428
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1556403

  SDS
alfa aesar™
A1556403
1g Each for $84.82

AAA1556414

  SDS
alfa aesar™
A1556414
25g Each for $1,506.76
Description & Specifications

Specifications

Formula Weight 213.16
Chemical Name or Material 3-(Trifluoromethyl)benzoylacetonitrile
Assay 97%
Molecular Formula C10H6F3NO
MDL Number MFCD00052315
CAS 27328-86-5
Beilstein 2373428
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 58° to 63°C
Quantity 1g

Applications
3-(Trifluoromethyl)benzoylacetonitrile is used as a pharmaceutical intermediate.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.