missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethyl)benzoylacetonitrile, 97%

Manufacturer:  Alfa Aesar™ A1556406

 View more versions of this product

Catalog No. AAA1556406



Description

Description

Applications
3-(Trifluoromethyl)benzoylacetonitrile is used as a pharmaceutical intermediate.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

3-(Trifluoromethyl)benzoylacetonitrile
5g
C10H6F3NO
2373428
GEPORLBYZLQDOB-UHFFFAOYSA-N
3-oxo-3-[3-(trifluoromethyl)phenyl]propanenitrile
520304
97%
58°C to 63°C
27328-86-5
MFCD00052315
3-trifluoromethylbenzoylacetonitrile, 3-oxo-3-3-trifluoromethyl phenyl propanenitrile, 3-oxo-3-3-trifluoromethyl phenyl propanenitrile, 3-trifluoromethyl benzoylacetonitrile, 2-cyano-3'-trifluoromethyl acetophenone, 3-oxo-3-3-trifluoromethyl-phenyl-propionitrile, pubchem12076, acmc-1cbhg, maybridge1_000123, 3-trifluoromethyl benzoyl acetonitrile
C1=CC(=CC(=C1)C(F)(F)F)C(=O)CC#N
213.159
213.16

Chemical Identifiers

27328-86-5
213.159
GEPORLBYZLQDOB-UHFFFAOYSA-N
520304
C1=CC(=CC(=C1)C(F)(F)F)C(=O)CC#N
C10H6F3NO
MFCD00052315
3-trifluoromethylbenzoylacetonitrile, 3-oxo-3-3-trifluoromethyl phenyl propanenitrile, 3-oxo-3-3-trifluoromethyl phenyl propanenitrile, 3-trifluoromethyl benzoylacetonitrile, 2-cyano-3'-trifluoromethyl acetophenone, 3-oxo-3-3-trifluoromethyl-phenyl-propionitrile, pubchem12076, acmc-1cbhg, maybridge1_000123, 3-trifluoromethyl benzoyl acetonitrile
3-oxo-3-[3-(trifluoromethyl)phenyl]propanenitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents