Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethyl)benzophenone, 97+%

$48.49 - $48.49

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethyl)benzophenone
CAS 728-81-4
Molecular Formula C14H9F3O
Formula Weight 250.22
MDL Number MFCD00000389
Beilstein 1971353
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2089706

 
alfa aesar™
B2089706
5g Each for $48.49
Description & Specifications

Specifications

Formula Weight 250.22
Chemical Name or Material 3-(Trifluoromethyl)benzophenone
Assay ≥97%
Molecular Formula C14H9F3O
MDL Number MFCD00000389
CAS 728-81-4
Beilstein 1971353
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)
Boiling Point 121° to 123°C (1.5mmHg)
Melting Point 51° to 53°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.