Please login to your online account to display your discounted pricing

3-(Trifluoromethyl)benzoic acid, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethyl)benzoic acid
Name Note 99%
CAS 454-92-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H5F3O2
Linear Formula CF3C6H4CO2H
Formula Weight 190.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC182540100

 
fisher scientific acros
182540100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Powder
Formula Weight 190.12
Quantity 10g
Color White to beige
Beilstein 09, 478
TSCA TSCA
EINECS Number 207-230-2
Melting Point 104° to 106°C
CAS Max % 100.0
Chemical Name or Material 3-(Trifluoromethyl)benzoic acid
Name Note 99%
Assay Percent Range 99%
Molecular Formula C8H5F3O2
Linear Formula CF3C6H4CO2H
MDL Number MFCD00002519
CAS 454-92-2
Infrared Spectrum Authentic
Synonym α, α, α-Trifluoro-m-toluic acid
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.