Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethyl)benzoic acid, 98+%

$65.09 - $797.08

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethyl)benzoic acid
CAS 454-92-2
Molecular Formula C8H5F3O2
Formula Weight 190.12
MDL Number MFCD00002519
Beilstein 2049239
Synonym alpha,alpha,alpha-Trifluoro-m-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1673709

 
alfa aesar™
A1673709
10g Each for $65.09

AAA1673718

 
alfa aesar™
A1673718
50g Each for $210.95

AAA1673730

 
alfa aesar™
A1673730
250g Each for $797.08
Description & Specifications

Specifications

Formula Weight 190.12
Melting Point 103° to 107°C
CAS 454-92-2
Quantity 10g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥98%
Boiling Point 237° to 240°C
MDL Number MFCD00002519
Chemical Name or Material 3-(Trifluoromethyl)benzoic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 2049239
Synonym alpha,alpha,alpha-Trifluoro-m-toluic acid
Molecular Formula C8H5F3O2
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.