Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethyl)benzhydrol, 97%

$69.00 - $69.00

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethyl)benzhydrol
CAS 728-80-3
Molecular Formula C14H11F3O
Formula Weight 252.23
MDL Number MFCD00014397
Beilstein 2418462
Synonym Phenyl 3-trifluoromethylphenyl carbinol; 3-(Trifluoromethyl)diphenylmethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1441403

  SDS
alfa aesar™
L1441403
1g Each for $69.00
Description & Specifications

Specifications

Formula Weight 252.23
Chemical Name or Material 3-(Trifluoromethyl)benzhydrol
CAS 728-80-3
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
MDL Number MFCD00014397
Synonym Phenyl 3-trifluoromethylphenyl carbinol; 3-(Trifluoromethyl)diphenylmethanol
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 2418462
Melting Point 62° to 64°C
Quantity 1g
Boiling Point 125° to 127°C (0.5mmHg)
Health Hazard 1 Warning
Molecular Formula C14H11F3O
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.