missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Trifluoromethyl)benzenesulfonyl Chloride, TCI America™

Manufacturer:  TCI America T19105G

 View more versions of this product

Catalog No. T19105G



Specifications

Specifications

3-(Trifluoromethyl)benzenesulfonyl Chloride
5g
C7H4ClF3O2S
3265
ONCAZCNPWWQQMW-UHFFFAOYSA-N
3-(trifluoromethyl)benzenesulfonyl chloride
2733250
Clear Liquid at 20°C
Yellow
777-44-6
MFCD00014724
3-trifluoromethyl benzenesulfonyl chloride, 3-trifluoromethyl benzene-1-sulfonyl chloride, m-trifluoromethylbenzenesulfonyl chloride, 3-trifluoromethyl benzenesulphonyl chloride, 3-trifluoromethylbenzenesulfochloride, 3-trifluoromethyl benzenesulfonylchloride, benzenesulfonyl chloride, 3-trifluoromethyl, 3-chlorosulphonyl benzotrifluoride, m-trifluoromethylphenylsulfonyl chloride, 3-trifluoromethyl-benzenesulfonyl chloride
C1=CC(=CC(=C1)S(=O)(=O)Cl)C(F)(F)F
244.612
244.61
≥98.0% (GC,T)

Chemical Identifiers

777-44-6
244.612
ONCAZCNPWWQQMW-UHFFFAOYSA-N
2733250
C1=CC(=CC(=C1)S(=O)(=O)Cl)C(F)(F)F
C7H4ClF3O2S
MFCD00014724
3-trifluoromethyl benzenesulfonyl chloride, 3-trifluoromethyl benzene-1-sulfonyl chloride, m-trifluoromethylbenzenesulfonyl chloride, 3-trifluoromethyl benzenesulphonyl chloride, 3-trifluoromethylbenzenesulfochloride, 3-trifluoromethyl benzenesulfonylchloride, benzenesulfonyl chloride, 3-trifluoromethyl, 3-chlorosulphonyl benzotrifluoride, m-trifluoromethylphenylsulfonyl chloride, 3-trifluoromethyl-benzenesulfonyl chloride
3-(trifluoromethyl)benzenesulfonyl chloride
Safety and Handling

Safety and Handling

TSCA : No

Documents