Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethyl)benzenesulfonyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethyl)benzenesulfonyl chloride
CAS 777-44-6
Molecular Formula C7H4ClF3O2S
Formula Weight 244.62
MDL Number MFCD00014724
UN Number UN3265
Beilstein 2650774
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1465706

  SDS
alfa aesar™
A1465706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1465714

  SDS
alfa aesar™
A1465714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 244.62
Chemical Name or Material 3-(Trifluoromethyl)benzenesulfonyl chloride
Assay 98%
Molecular Formula C7H4ClF3O2S
MDL Number MFCD00014724
CAS 777-44-6
UN Number UN3265
Beilstein 2650774
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Flash Point >110°C (230°F)
Boiling Point 79° to 80°C (1mmHg)
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.