missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethyl)benzenesulfonyl chloride, 98%

Manufacturer:  Alfa Aesar™ A1465703

 View more versions of this product

Catalog No. AAA1465703



Specifications

Specifications

3-(Trifluoromethyl)benzenesulfonyl chloride
79°C to 80°C (1mmHg)
777-44-6
MFCD00014724
2650774
ONCAZCNPWWQQMW-UHFFFAOYSA-N
3-(trifluoromethyl)benzenesulfonyl chloride
2733250
98%
>110°C (230°F)
1g
C7H4ClF3O2S
UN3265
3-trifluoromethyl benzenesulfonyl chloride, 3-trifluoromethyl benzene-1-sulfonyl chloride, m-trifluoromethylbenzenesulfonyl chloride, 3-trifluoromethyl benzenesulphonyl chloride, 3-trifluoromethylbenzenesulfochloride, 3-trifluoromethyl benzenesulfonylchloride, benzenesulfonyl chloride, 3-trifluoromethyl, 3-chlorosulphonyl benzotrifluoride, m-trifluoromethylphenylsulfonyl chloride, 3-trifluoromethyl-benzenesulfonyl chloride
C1=CC(=CC(=C1)S(=O)(=O)Cl)C(F)(F)F
244.612
244.62

Chemical Identifiers

777-44-6
244.612
ONCAZCNPWWQQMW-UHFFFAOYSA-N
2733250
C1=CC(=CC(=C1)S(=O)(=O)Cl)C(F)(F)F
C7H4ClF3O2S
MFCD00014724
3-trifluoromethyl benzenesulfonyl chloride, 3-trifluoromethyl benzene-1-sulfonyl chloride, m-trifluoromethylbenzenesulfonyl chloride, 3-trifluoromethyl benzenesulphonyl chloride, 3-trifluoromethylbenzenesulfochloride, 3-trifluoromethyl benzenesulfonylchloride, benzenesulfonyl chloride, 3-trifluoromethyl, 3-chlorosulphonyl benzotrifluoride, m-trifluoromethylphenylsulfonyl chloride, 3-trifluoromethyl-benzenesulfonyl chloride
3-(trifluoromethyl)benzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Documents