Learn More
3-(Trifluoromethyl)benzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1465706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-(Trifluoromethyl)benzenesulfonyl chloride | |
1.526 | |
>110°C (230°F) | |
1.487 | |
5 g | |
2650774 | |
3-trifluoromethyl benzenesulfonyl chloride, 3-trifluoromethyl benzene-1-sulfonyl chloride, m-trifluoromethylbenzenesulfonyl chloride, 3-trifluoromethyl benzenesulphonyl chloride, 3-trifluoromethylbenzenesulfochloride, 3-trifluoromethyl benzenesulfonylchloride, benzenesulfonyl chloride, 3-trifluoromethyl, 3-chlorosulphonyl benzotrifluoride, m-trifluoromethylphenylsulfonyl chloride, 3-trifluoromethyl-benzenesulfonyl chloride | |
C1=CC(=CC(=C1)S(=O)(=O)Cl)C(F)(F)F | |
244.612 | |
244.62 |
777-44-6 | |
79°C to 80°C (1 mmHg) | |
C7H4ClF3O2S | |
MFCD00014724 | |
UN3265 | |
Moisture sensitive | |
ONCAZCNPWWQQMW-UHFFFAOYSA-N | |
3-(trifluoromethyl)benzenesulfonyl chloride | |
2733250 | |
98% |
Chemical Identifiers
777-44-6 | |
244.612 | |
ONCAZCNPWWQQMW-UHFFFAOYSA-N | |
2733250 | |
C1=CC(=CC(=C1)S(=O)(=O)Cl)C(F)(F)F |
C7H4ClF3O2S | |
MFCD00014724 | |
3-trifluoromethyl benzenesulfonyl chloride, 3-trifluoromethyl benzene-1-sulfonyl chloride, m-trifluoromethylbenzenesulfonyl chloride, 3-trifluoromethyl benzenesulphonyl chloride, 3-trifluoromethylbenzenesulfochloride, 3-trifluoromethyl benzenesulfonylchloride, benzenesulfonyl chloride, 3-trifluoromethyl, 3-chlorosulphonyl benzotrifluoride, m-trifluoromethylphenylsulfonyl chloride, 3-trifluoromethyl-benzenesulfonyl chloride | |
3-(trifluoromethyl)benzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only