Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethyl)benzenesulfonamide, 97%

$88.97 - $321.72

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethyl)benzenesulfonamide
CAS 672-58-2
Molecular Formula C7H6F3NO2S
Formula Weight 225.19
MDL Number MFCD00042421
Beilstein 2696480
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1971503

  SDS
alfa aesar™
L1971503
1g Each for $88.97

AAL1971506

  SDS
alfa aesar™
L1971506
5g Each for $321.72
Description & Specifications

Specifications

Formula Weight 225.19
Molecular Formula C7H6F3NO2S
MDL Number MFCD00042421
CAS 672-58-2
Chemical Name or Material 3-(Trifluoromethyl)benzenesulfonamide
Assay 97%
Beilstein 2696480
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 122° to 126°C
Quantity 1g
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.