Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Trifluoromethyl)benzamide, 98%

$295.32 - $295.32

Chemical Identifiers

Chemical Name or Material 3-(Trifluoromethyl)benzamide
CAS 1801-10-1
Molecular Formula C8H6F3NO
Formula Weight 189.14
MDL Number MFCD00014802
Beilstein 1955599
Synonym alpha,alpha,alpha-Trifluoro-m-toluamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2004914

  SDS
alfa aesar™
B2004914
25g Each for $295.32

AAB2004906

  SDS
alfa aesar™
B2004906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 3-(Trifluoromethyl)benzamide
Melting Point 122° to 126°C
Quantity 25g
CAS 1801-10-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Molecular Formula C8H6F3NO
Formula Weight 189.14
MDL Number MFCD00014802
Beilstein 1955599
Synonym alpha,alpha,alpha-Trifluoro-m-toluamide
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.